Orders LinkContactUsLinkComponent
Compression Union Tees, PFA
Compression Union Tees, PFA
Supplier:   VWR International
Compression Union Tees, PFA
Supplier:  VWR International
Certificates
Compression Union Tees, PFA
1 Options Available Below
Product Details & Documents
Get a secure seal at higher pressures.
  • Constructed of durable PFA
Compression fittings are an excellent choice to connect rigid tubing systems and withstand higher pressures than barbed fittings. Assemble the fitting to the tubing by sliding the tubing through a sealing ferrule and using the nut to compress down to form a secure seal.
Specifications for Product Family
Specifications
  • Sterilization:
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)
  • Warranty:
  • 18 months
  • 18 months
  • 18 months
  • 18 months
  • 18 months
  • 1 year
Specifications
Product Family Options
Product Information
  • Connection Type
    Flow pattern
    Compression Size / Tube O.D. (Port 3)
    Fitting Material
    Availability
    Price
  • MFLX31320-29
    Compression Union Tee, PFA, ⅛"; 1/EA
    Compression Union Tee, PFA, ⅛"; 1/EA
    Compression Union
    T
    ⅛"
    PFA
    Discontinued
    Specifications:
    CE certificate
    More
    Specifications
    Cat. No.
    MFLX31320-29
    Sterilization
    Autoclave or ethylene oxide (ETO)
    Warranty
    18 months
    CE certificate
  • MFLX31320-30
    Compression Union Tee, PFA, ¼"; 1/EA
    Compression Union Tee, PFA, ¼"; 1/EA
    Compression Union
    T
    ¼"
    PFA
    Discontinued
    Specifications:
    CE certificate
    More
    Specifications
    Cat. No.
    MFLX31320-30
    Sterilization
    Autoclave or ethylene oxide (ETO)
    Warranty
    18 months
    CE certificate
  • MFLX31320-31
    Compression Union Tee, PFA, ⅜"; 1/EA
    Compression Union Tee, PFA, ⅜"; 1/EA
    Compression Union
    T
    ⅜"
    PFA
    Discontinued
    Specifications:
    CE certificate
    More
    Specifications
    Cat. No.
    MFLX31320-31
    Sterilization
    Autoclave or ethylene oxide (ETO)
    Warranty
    18 months
    CE certificate
  • MFLX31320-32
    Compression Union Tee, PFA, ½"; 1/EA
    Compression Union Tee, PFA, ½"; 1/EA
    Compression Union
    T
    ½"
    PFA
    Discontinued
    Specifications:
    CE certificate
    More
    Specifications
    Cat. No.
    MFLX31320-32
    Sterilization
    Autoclave or ethylene oxide (ETO)
    Warranty
    18 months
    CE certificate
  • MFLX31320-33
    Compression Union Tee, PFA, ¾"; 1/EA
    Compression Union Tee, PFA, ¾"; 1/EA
    Compression Union
    T
    ¾"
    PFA
    Discontinued
    Specifications:
    CE certificate
    More
    Specifications
    Cat. No.
    MFLX31320-33
    Sterilization
    Autoclave or ethylene oxide (ETO)
    Warranty
    18 months
    CE certificate
  • MFLX31321-48
    Compression Union Tee, PFA, 8mm; 1/EA
    Compression Union Tee, PFA, 8mm; 1/EA
    Compression Union
    T
    8 mm
    PFA
    Log in for availability
    Log in for availability
    Each
    Each - $214.75
    $214.75
    Each
       $214.75
    Specifications:
    CE certificate
    More
    Specifications
    Cat. No.
    MFLX31321-48
    Sterilization
    Autoclave or ethylene oxide (ETO)
    Warranty
    1 year
    CE certificate