Noryl® Corrosion Resistant Chemical-Resistance Centrifugal Pumps
Supplier: Avantor
Certificates
About this item
Chemical-resistant pump heads and impellers handle a variety of corrosive and caustic fluids over the entire pH range.
- Ideal for circulation, transfer, and chemical feed applications
- NEMA 56 C-face motors feature thermal overload protection
These Noryl® corrosion resistant chemical-resistance centrifugal pumps are ideal for circulation, transfer, and chemical feed applications. Pumps are constructed with a Noryl pump head and impeller, carbon/ceramic face seal, Noryl seal encasement, and Viton® O-ring.
Delivery: Includes all pumps come with stainless steel plated hardware and conduit box for hardwiring.
- Environmentally Preferable:
Frequently Bought Together