Chemical-Duty Self-Priming Transfer Pumps
Supplier: MARCH MANUFACTURING INC
Certificates
About this item
Different wetted materials for different chemical compatibilities.
- Pumps with glass-filled polypropylene heads are ideal for transferring a wide range of caustic materials
- Pumps with virgin PVDF heads are ideal for DI H2O, strong acids, and halogens
Self-priming pumps with glass-filled polypropylene heads are capable of lifting water up to 10 feet.
Delivery: Includes conduit box for hardwiring.
- Environmentally Preferable:
Frequently Bought Together