Chem-Tec Adjustable Mini Low-Flow Switches
: Avantor
Adjustable set points trigger SPST contacts rated for 70 VA.
- Choose from brass or TFE materials
Mini Low-Flow Switches are configured to open the SPST contact when flow goes beyond the set point or stops. Typical applications include gas/liquid sampling; chemical injection; pollution control monitoring; atmospheric furnaces; and process systems. Choose from a brass switch (32929-00) for pressures up to 1500 psi or a switch made from PTFE (32929-02) for good chemical compatibility. Wetted materials are brass, epoxy, and Viton® for model 32929-00; PTFE for model 32929-02. Repeatability is ±2%. Measure 1-¼"W x 2-5/8"H x ¾"D.
- Environmentally Preferable:
Frequently Bought Together