Hayward® True Union Ball Check Valves with Foot Valve Screens
: Avantor
Unique square-cut elastomer seat provides seal at very low back pressures.
- Suitable for use with Hayward® True Union Ball Check Valves
Quickly and easily install either vertically or horizontally. These valves are assembled with water soluble, silicone-free lubricant and constructed of a one-piece PVC body.
The foot valve screen helps prevent unwanted material from entering the system.
- Environmentally Preferable:
Frequently Bought Together