Parker Hannifin British Standard Pipe Threaded (BSPP) Adapters
Supplier: Avantor
Certificates
About this item
Convert NPT-threaded systems to BSPP threads—or visa versa—with these stainless steel fittings.
- Constructed of durable stainless steel
Use BSP parallel (BSPP) fittings for connections where pressure-tight joints are not made on the thread (as in connections with peripheral seals). Stainless steel fittings are chemically resistant.
- Environmentally Preferable:
Frequently Bought Together