Order Entry
United States
ContactUsLinkComponent
TCEP-HCl (Tris(2-carboxyethyl)phosphine hydrochloride), No-Weigh™ Format, Pierce™
Corrosive
TCEP-HCl (Tris(2-carboxyethyl)phosphine hydrochloride), No-Weigh™ Format, Pierce™
Catalog # 103523-932
Supplier:  Invitrogen
CAS Number:  
Corrosive
TCEP-HCl (Tris(2-carboxyethyl)phosphine hydrochloride), No-Weigh™ Format, Pierce™
Catalog # 103523-932
Supplier:  Invitrogen
Supplier Number:  A35349
CAS Number:  

Specifications

  • Size:
    10 x 1 mg
  • Cat. No.:
    103523-932

Specifications

About this item

 :
 :286.65 g/mol
Storage Temperature:Refrigerator
UN:3261
ADR:8,II
 :GHS05
CAS Number:51805-45-9
Formula:C₉H₁₆ClO₆P
MDL Number:MFCD00145469