Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Antibody Type:Primary
- Antigen Name:Ectonucleotide pyrophosphatase/phosphodiesterase family member 1
- Antigen Symbol:E-NPP 1
- Clonality:Polyclonal
- Clone:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Sheep
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:0.1 mg
- Form:The antibody is lyophilized in 0.05 M phosphate buffer, 0.1 M NaCl, pH 7.2. AZIDE FREE.
- Antigen Synonyms:Phosphodiesterase I/nucleotide pyrophosphatase 1|NPP-1|Ectonucleotide pyrophosphatase/phosphodiesterase family member 1|PC-1|E-NPP 1|Membrane component chromosome 6 surface marker 1|Plasma-cell membrane glycoprotein
- Storage Buffer:Add 0.2 ml of deionized water and let the lyophilized pellet dissolve completely. Slight turbidity may occur after reconstitution, which does not affect activity of the antibody. In this case clarify the solution by centrifugation.
- Shipping Temperature:At ambient temperature
- Purification:Immunoaffinity chromatography on a column with immobilized recombinant Human ENPP1 Protein.
- Cat. No.:102830-258
- Supplier no.:RD184124100
Quality control test, Indirect ELISA – to determine titer of the antibody , SDS PAGE – to determine purity of the antibody, and BCA - to determine quantity of the antibody.
Research topic. Bone and cartilage metabolism, Diabetology - Other Relevant Products, Energy metabolism and body weight regulation
Storage temperature: The lyophilized antibody remains stable and fully active until the expiry date when stored at –20 °C. Aliquot the product after reconstitution to avoid repeated freezing/thawing cycles and store frozen at –80 °C. Reconstituted antibody can be stored at 4 °C for a limited period of time; it does not show decline in activity after one week at 4 °C.