Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Name:Trefoil Factor 3
- Antigen Symbol:TFF3
- Clonality:Polyclonal
- Clone:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Sheep
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:0.1 mg
- Form:The antibody is lyophilized in 0.05 M phosphate buffer, 0.1 M NaCl, pH 7.2. AZIDE FREE.
- Antigen Synonyms:TFF3|ITF|Polypeptide P1.B|TFI|Intestinal trefoil factor|P1.B
- Storage Buffer:Add 0.2 ml of deionized water and let the lyophilized pellet dissolve completely. Slight turbidity may occur after reconstitution, which does not affect activity of the antibody. In this case clarify the solution by centrifugation.
- Shipping Temperature:At ambient temperature
- Purification:Immunoaffinity chromatography on a column with immobilized recombinant Human Trefoil Factor 3.
- Cat. No.:101076-692
- Supplier no.:RD184160100
Specifications
About this item
Quality control test, Indirect ELISA – to determine titer of the antibody , SDS PAGE – to determine purity of the antibody, and BCA - to determine quantity of the antibody.
Trefoil factor 3 (TFF3, also known as intestinal trefoil factor) belongs together with TFF1 and TFF2 to a small group of mucin-associated peptides. TFF3 contains seven cysteine residues, six of which form disulfide bonds to create a characteristic three-leafed structure. Due to its compact structure, TFF3 is extremely resistant toward acids, proteolytical cleavage or heat degradation. Monomeric form of TFF3 consists of 60 amino acids and has 6.7 kDa, while the dimer (13.1 kDa) consists of 118 amino acids.
Storage temperature: The lyophilized antibody remains stable and fully active until the expiry date when stored at –20 °C. Aliquot the product after reconstitution to avoid repeated freezing/thawing cycles and store frozen at –80 °C. Reconstituted antibody can be stored at 4 °C for a limited period of time; it does not show decline in activity after one week at 4 °C.