Sircol™ Soluble and Insoluble Collagen, BioVendor
Collagen is the most abundant protein found in animals. During a healthy life span the insoluble, covalent, cross-linked collagen fibers retain their biophysical functions and shapes. The collagen fibers remain isolated from most of the biochemical activities of the cells.
- Dye-binding method
- It replaces the conventional hydroxyproline analysis. No more working with concentrated acids or alkalis
- The Sircol Insoluble Assay uses mild acid and temperature treatment for 2 to 3 hrs
- Storage Conditions:All components are stable for one year, (from Invoice Date), when stored at 15–25°C. The glass vial containing collagen standard should be stored at +4°C once opened.
- Detection Limit:10 μg
Frequently Bought Together