Order Entry
United States
ContactUsLinkComponent
ENDONEXT™ EndoZyme® II Go Fast Recombinant Factor C (rFC) Endotoxin Detection Assay, BioVendor
ENDONEXT™ EndoZyme® II Go Fast Recombinant Factor C (rFC) Endotoxin Detection Assay, BioVendor
Catalog # 103739-724
Supplier:  BioVendor
CAS Number:  
ENDONEXT™ EndoZyme® II Go Fast Recombinant Factor C (rFC) Endotoxin Detection Assay, BioVendor
Catalog # 103739-724
Supplier:  BioVendor
Supplier Number:  890031
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    ENDONEXT™ EndoZyme® II Go Fast rFC endotoxin detection assay
  • Size:
    1 Kit
  • Storage Conditions:
    All components are stable for one year, (from invoice date), when stored at 15…25 ºC. Do not store below 4 ºC. Once opened the glass vial containing sGAG standard should however be stored at 4 ºC.
  • Cat. No.:
    103739-724
  • No. of tests:
    192

Specifications

About this item

The ENDONEXT™ EndoZyme® II assay is the enhanced second generation of EndoZyme®, based on recombinant Factor C (rFC), the synthetic analogue to the native endotoxin-specific principal receptor in the limulus amebocyte lysate (LAL) cascade.

  • Assay run time: 20 minutes (0.05 EU/ml), 60 minutes (0.005 EU/ml)
  • Preloaded microplate eliminates manual preparation of standard dilutions and PPCs
  • Over 50% reduction in handling time compared to conventional microplate assays
  • Significantly reduces risk of handling error during preparation steps
  • Higher precision and rate of valid results ideal for in-process control of water and raw materials, as well as product release testing ideal for automation and parallel processing of plates

Instead of being based on an animal source, EndoZyme® II is a sustainable alternative method in line with regulatory requirements.

The rapid GO version of EndoZyme® II features GOPLATE™ - a microplate pre-filled with dried control standard endotoxin, standard curve 0.005 - 50 EU/ml and positive product Ccontrols (PPC) 0.5 EU/ml, all in duplicate replicates fulfilling pharmacopoeia standard curve requirements.

This new version of EndoZyme® enables endotoxin testing in pharmaceutical grade water, injectable drugs and other pharmaceutical products. EndoZyme® II GO allows an easy and fast workflow, and is particularly adapted to raw materials and final product testing.

Ships at ambient temperature. Upon receipt, store the product at the temperature recommended below. Unopened kits are stable at 2...8°C until the expiry date printed on the label. The calibration range is 0.005 – 50 EU/ml. The limit of detection is 0.005 EU/ml.