Helicobacter Pylori Neutrophil-activating Protein A Helicobacter Pylori E. coli, 0.1 mg
Catalog # 103739-592
Supplier: BioVendor
CAS Number:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Helicobacter pylori
- Size:0.1 mg
- Storage Conditions:Store the lyophilized protein at –80 °C. Lyophilized protein remains stable until the expiry date when stored at –80 °C. Aliquot reconstituted protein to avoid repeated freezing/thawing cycles and store at –80 °C for long term storage. Reconstituted protein can be stored at 4 °C for a week
- Endotoxin Content:<0.1 EU/μg
- Reconstitution Instructions:Add 200 µl of deionized water to prepare a working stock solution of approximately 0.5 mg/ml and let the lyophilized pellet dissolve completely. Filter sterilize your culture media/working solutions containing this non-sterile product before using in cell culture.
- Protein Synonyms:HP-NAP|DNA protection during starvation protein|Bacterioferritin
- Protein/Peptide Name:Helicobacter pylori Neutrophil-activating Protein A Helicobacter pylori E. coli
- Purity:>95%
- Molecular Weight:18.2 kDa (calculated)
- Sequence:MKHHHHHHASMKTFEILKHLQADAIVLFMKVHNFHWNVKGTDFFNVHKATEEIYEEFADMFDDLAERIVQLGHHPLVTLSEAIKLTRVKEETKTSFHSKDIFKEILEDYKYLEKEFKELSNTAEKEGDKVTVTYADDQLAKLQKSIWMLQAHLA
- Endotoxin Level:<0.1 EU/μg
- Formulation:Filtered (0.4 μm) and lyophilized in 20 mM Tris buffer, 50 mM NaCl, pH 7.5
- Shipping Temperature:At ambient temperature
- Tested Applications:Western blotting, ELISA
- Cat. No.:103739-592
Specifications
About this item
Total 154 AA. MW: 18.2 kDa (calculated). UniProtKB acc.no. P43313 (Met1-Ala144). N-terminal His-tag (10 extra AA). Protein identity confirmed by LC-MS/MS.
- Quality control test
- BCA to determine quantity of the protein
- SDS PAGE to determine purity of the protein
- LAL to determine quantity of endotoxin
Caution: Research use only.