CD5 Antigen-Like Human HEK293, 0.1 mg
103739-678
:
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Size:0.1 mg
- Storage Conditions:Store the lyophilized protein at –80 °C. Lyophilized protein remains stable until the expiry date when stored at –80 °C. Aliquot reconstituted protein to avoid repeated freezing/thawing cycles and store at –80 °C for long term storage. Reconstituted protein can be stored at 4 °C for a week
- Endotoxin Content:<1.0 EU/µg
- Reconstitution Instructions:Add 200 µl of deionized water to prepare a working stock solution of approximately 0.5 mg/ml and let the lyophilized pellet dissolve completely. Filter sterilize your culture media/working solutions containing this non-sterile product before using in cell culture.
- Protein Synonyms:CD5L|UNQ203/PRO229|CT-2|AIM|SP-alpha|IgM-associated peptide|Apoptosis inhibitor expressed by macrophages|API6
- Protein/Peptide Name:CD5 Antigen-like human HEK293
- Purity:>95%
- Molecular Weight:36.9 kDa (calculated)
- Sequence:SPSGVRLVGGLHRCEGRVEVEQKGQWGTVCDDGWDIKDVAVLCRELGCGAASGTPSGILYEPPAEKEQKVLIQSVSCTGTEDTLAQCEQEEVYDCSHDEDAGASCENPESSFSPVPEGVRLADGPGHCKGRVEVKHQNQWYTVCQTGWSLRAAKVVCRQLGCGRAVLTQKRCNKHAYGRKPIWLSQMSCSGREATLQDCPSGPWGKNTCNHDEDTWVECEDPFDLRLVGGDNLCSGRLEVLHKGVWGSVCDDNWGEKEDQVVCKQLGCGKSLSPSFRDRKCYGPGVGRIWLDNVRCSGEEQSLEQCQHRFWGFHDCTHQEDVAVICSGHHHHHH
- Endotoxin Level:<1.0 EU/µg
- Formulation:Filtered (0.4 μm) and lyophilized in phosphate buffered saline pH 7.5
- Shipping Temperature:At ambient temperature
- Tested Applications:Western blotting, ELISA, Cell culture and/or animal studies
- Cat. No.:103739-678
Total 334 AA. MW: 36.9 kDa (calculated). UniProtKB acc. No. O43866 (Ser20–Gly347). C-terminal His-tag (6 extra AA). Protein identity confirmed by LC-MS/MS.
- Quality control test
- BCA to determine quantity of the protein
- SDS PAGE to determine purity of the protein
- LAL to determine quantity of endotoxin
: Research use only.