KIM-1 Human HEK293
Catalog # 103739-798
Supplier: BioVendor
CAS Number:
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Size:0.1 mg
- Storage Conditions:Store the lyophilized protein at –80 °C. Lyophilized protein remains stable until the expiry date when stored at –80 °C. Aliquot reconstituted protein to avoid repeated freezing/thawing cycles and store at –80 °C for long term storage. Reconstituted protein can be stored at 4 °C for a week
- Endotoxin Content:<1.0 EU/µg
- Reconstitution Instructions:Add 200 µl of deionized water to prepare a working stock solution of approximately 0.5 mg/ml and let the lyophilized pellet dissolve completely. Filter sterilize your culture media/working solutions containing this non-sterile product before using in cell culture.
- Protein Synonyms:TIM-1|KIM-1|T-cell immunoglobulin mucin receptor 1|HAVCR1|Hepatitis A Virus Cellular Receptor 1|Kidney injury molecule 1|T-cell immunoglobulin and mucin domain-containing protein 1|T-cell membrane protein 1|HAVcr-1|TIMD-1|TIM
- Protein/Peptide Name:KIM-1 Human HEK293
- Purity:>90%
- Molecular Weight:30.5 kDa (calculated)
- Sequence:ASSVKVGGEAGPSVTLPCHYSGAVTSMCWNRGSCSLFTCQNGIVWTNGTHVTYRKDTRYKLLGDLSRRDVSLTIENTAVSDSGVYCCRVEHRGWFNDMKITVSLEIVPPKVTTTPIVTTVPTVTTVRTSTTVPTTTTVPMTTVPTTTVPTTMSIPTTTTVLTTMTVSTTTSVPTTTSIPTTTSVPVTTTVSTFVPPMPLPRQNHEPVATSPSSPQPAETHPTTLQGAIRREPTSSPLYSYTTDGNDTVTESSDGLWNNNQTQLFLEHSLLTANTTKLHHHHHH
- Endotoxin Level:<1.0 EU/µg
- Formulation:Filtered (0.4 μm) and lyophilized from 0.5 mg/ml solution in phosphate buffered saline, 5% w/v trehalose
- Shipping Temperature:At ambient temperature
- Tested Applications:Western blotting, ELISA, Cell culture and/or animal studies
- Cat. No.:103739-798
Specifications
About this item
Total 283 AA. MW: 30.5 kDa (calculated). UniProtKB acc. No. Q96D42 (Ser21–Thr288). N-terminal linker (2 extra AA), C-terminal linker (2 extra AA) and C-terminal His-tag (6 extra AA). Protein identity confirmed by LC-MS/MS.
- Quality control test
- BCA to determine quantity of the protein
- SDS PAGE to determine purity of the protein
- LAL to determine quantity of endotoxin
Caution: Research use only.