Replacement Tips for Tweezers, Excelta
Catalog # 10066-504
Supplier: Excelta
Specifications
- Color:Black
- Description:Carbon fiber tips, straight, for 116-CF-RTX tweezers
- Environmentally Preferable:
- Cat. No.:10066-504
- Supplier no.:116-CF-RTX
Specifications
About this item
Set of 2 Carbofib™ replacement tips for 116-CF SMD straight tweezers.
Tips have a .039" (1mm) hole. This tool is conductive (10² ohms/sq).