Order Entry
United States
ContactUsLinkComponent
KMUH (N-(κ-maleimidoundecanoic acid) hydrazide trifluoroacetic acid salt), Pierce™
KMUH (N-(κ-maleimidoundecanoic acid) hydrazide trifluoroacetic acid salt), Pierce™
Catalog # PI22111
Supplier:  Invitrogen
CAS Number:  
undefined
KMUH (N-(κ-maleimidoundecanoic acid) hydrazide trifluoroacetic acid salt), Pierce™
Catalog # PI22111
Supplier:  Invitrogen
Supplier Number:  22111
CAS Number:  

Specifications

  • Size:
    50 mg
  • Cat. No.:
    PI22111
  • Supplier no.:
    22111

Specifications

About this item

 :
 :409.40 g/mol
 :GHS07
Storage Temperature:Refrigerator
CAS Number:1593661-78-9
 :359436-62-7
Formula:C₁₅H₂₅N₃O₃·CF₃CO₂H