Order Entry
United States
Orders LinkContactUsLinkComponent
3-(4-Bromophenyl)propionic acid 97%

3-(4-Bromophenyl)propionic acid 97%

Irritant
3-(4-Bromophenyl)propionic acid 97%
Certificates
3-(4-Bromophenyl)propionic acid 97%

2 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
  • CAS Number:1643-30-7
  • Formula:C₉H₉BrO₂
  • MDL Number:MFCD01310793
  • Molecular Weight:229.07
  • Melting Pt:133…136 °C
  • Storage Temperature:Ambient
  • UN:3335
  • ADR:9,III
Specifications for Product Family

Specification Test Results

  • Appearance:
    White to yellow to beige Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    134 to 138 °C
  • HPLC:
    ≥96.0 %
Product Family Options

Product Information

  • Pack type
    Size
    Availability
    Price
  • Glass bottle
    1 g
    Log in for availability
    Log in for availability
    Each (1g)
    Each (1g) - $37.30
    $37.30
    Each (1g)
       $37.30

    Supplier Number: 39945-0010
  • Glass bottle
    5 g
    Log in for availability
    Log in for availability
    Each (5g)
    Each (5g) - $103.00
    $103.00
    Each (5g)
       $103.00

    Supplier Number: 39945-0050

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.