
Lactic acid ≥85%
Catalog # TS41296-5000
Supplier: Thermo Scientific Chemicals
CAS Number:
Specifications
- Pack type:Bottle
- Size:500 g
- Cat. No.:TS41296-5000
Specifications
Specification Test Results
- Appearance:Clear colorless Liquid
- Residue after ignition:≤0.02 %
- Titration excess NaOH:85.0 to 90.0 %
- Substance darkened by H2SO4:Passes test
- Chloride (Cl):≤10 ppm
- Sulfate (SO4):≤20 ppm
- Iron (Fe):≤5 ppm
- Heavy metals (as Pb):≤5 ppm
Specification Test Results
About this item
:
Merck Index:13,05351
Boiling Pt:122 °C (20 hPa)
Melting Pt:16.8 °C
Density:1.357 g/cm³ (20 °C)
Flash Pt:113 °C (closed cup)
:90.08 g/mol
Storage Temperature:Ambient
EINECS:209-954-4
:GHS05
CAS Number:50-21-5
:598-82-3
Formula:H₃CCH(OH)COOH
MDL Number:MFCD00004520