
4-Chlorocinnamic acid predominantly trans 99%
Supplier: Thermo Scientific Chemicals
Certificates
About this item
CAS Number:1615-02-7
Formula:C₉H₇ClO₂
MDL Number:MFCD00004396
:182.61
Boiling Pt:311 °C (1013 hPa)
Melting Pt:246…250 °C
EINECS:216-564-8
:GHS07
Specification Test Results
- Appearance:White Crystalline powder
- Infrared spectrum:Conforms
- Melting point:246 to 250 °C
- Titration with NaOH:98.5 to 101.5 %
- UV:at 230 nm A: ≤1
- Color scale:≤10 APHA
- Residue after evaporation:≤0.001 %
- Carbonyl compounds:≤0.01 % Butyraldehyde
- Titrable acid:≤0.0005 meq/g
Specification Test Results
Frequently Bought Together