ContactUsLinkComponent
Oxidation-Reduction of Complex Ions
Oxidation-Reduction of Complex Ions
Catalog # 470338-586
Supplier:  Ward's Science
undefined
Oxidation-Reduction of Complex Ions
Catalog # 470338-586
Supplier:  Ward's Science
Supplier Number:  IS7030
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Subjects:
    HS-PS1-2, MS-PS1-2
  • Description:
    Oxidation-reduction of complex ions
  • Cat. No.:
    470338-586
  • Includes:
    Instructional manual and SDS booklet
  • No. of uses:
    5
Specifications
About this item
Observe color changes through chemical redox reactions of complex ions.
  • Understand and demonstrate redox reactions of complex ions
  • Aligned to NGSS practices
  • Cross cutting concepts: patterns
  • Disciplinary core ideas: PS1.A, PS1.B
  • Learn engineering practices: analyzing and interpreting data
Hydrogen peroxide is added to a solution of sodium potassium tartrate and heated. Copper sulfate solution is added and the solution turns light blue. With continued heating the solution foams and turns orange-gold.
Caution: WARNING: This product can expose you to chemicals including strong inorganic acid mists containing sulfuric acid, which are known to the state of California to cause cancer. For more information go to www.P65Warnings.ca.gov