ContactUsLinkComponent
Oscillating Reactions - The REDOX Rainbow
Oscillating Reactions - The REDOX Rainbow
Catalog # 470338-580
Supplier:  Ward's Science
undefined
Oscillating Reactions - The REDOX Rainbow
Catalog # 470338-580
Supplier:  Ward's Science
Supplier Number:  IS7027
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Subjects:
    MS-PS1-2
  • Description:
    Oscillating reactions - the redox rainbow
  • Cat. No.:
    470338-580
  • Includes:
    Instructional manual and SDS booklet
  • No. of uses:
    5
Specifications
About this item
Perform an oscillating redox reaction with hydrogen peroxide, potassium iodide, and potassium permanganate.
  • Understand and demonstrate oscillating redox reactions
  • Aligned to NGSS practices
  • Cross cutting concepts: patterns
  • Disciplinary core ideas: PS1.A, PS1.B
  • Learn engineering practices: analyzing and interpreting data
For this demonstration, add hydrogen peroxide to a colorless solution of potassium iodide to produce a red color, or you can add the hydrogen peroxide to a purple solution of potassium permanganate to create a colorless solution.
Caution: WARNING: This product can expose you to chemicals including strong inorganic acid mists containing sulfuric acid, which are known to the state of California to cause cancer. For more information go to www.P65Warnings.ca.gov