Specifications
- Subjects:HS-PS1-2. MS-PS1-2
- Description:Thionin and iron: a light induced redox reaction
- Cat. No.:470338-568
- Includes:Instructional manual and SDS booklet
- No. of uses:5
Specifications
About this item
Understand the effect of light energy on a redox reaction.
- Understand and demonstrate redox reactions
- Aligned to NGSS practices
- Cross cutting concepts: patterns
- Disciplinary core ideas: PS1.A, PS1.B
- Learn engineering practices: analyzing and interpreting data
In this lab a thionin-iron solution is prepared. Exposing the solution to light energy causes the colored thionin to be reduced by iron (II) ions, resulting in a clear solution. When half of the solution, in a single container, is exposed to light it will turn clear while the half not exposed to light will remain colored.