Specifications
- Subjects:MS-PS1-3
- Description:Forming red, white, and blue
- Cat. No.:470338-566
- Includes:Instructional manual and SDS booklet
- No. of uses:5
Specifications
About this item
Introduce different chemical reaction types through a colorful demonstration.
- Understand and demonstrate different chemical reaction types
- Aligned to NGSS practices
- Cross cutting concepts: patterns
- Disciplinary core ideas: PS1.A, PS1.B
- Learn engineering practices: analyzing and interpreting data
Introduce chemical reaction types. This is a patriotic demonstration that illustrates complex ion formation, double replacement while introducing chemical reaction types.