Specifications
- Subjects:MS-PS1-2
- Description:Formation of silicate (red and white precipitates)
- Cat. No.:470338-562
- Includes:Instructional manual and SDS booklet
- No. of uses:5
Specifications
About this item
Show how silicates of divalent ions are insoluble in water whereas silicates of monovalent ions are soluble.
- Understand and demonstrate the differences between divalent and monovalent ions through the formation of a silicate
- Aligned to NGSS practices
- Cross cutting concepts: patterns
- Disciplinary core ideas: PS1.A, PS1.B
- Learn engineering practices: analyzing and interpreting data