ContactUsLinkComponent
Ampholytes Chemical Demo
Ampholytes Chemical Demo
Catalog # 470338-556
Supplier:  Ward's Science
undefined
Ampholytes Chemical Demo
Catalog # 470338-556
Supplier:  Ward's Science
Supplier Number:  IS7003
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Subjects:
    MS-PS1-2
  • Description:
    Ampholytes chemical demo
  • Cat. No.:
    470338-556
  • Includes:
    Instructional manual and SDS booklet
  • No. of uses:
    5
Specifications
About this item
Observe the reaction of an amplolyte with both an acid and a base.
  • Understand and demonstrate ampholyte compounds
  • Aligned to NGSS practices
  • Cross cutting concepts: patterns
  • Disciplinary core ideas: PS1.A, PS1.B
  • Learn engineering practices: analyzing and interpreting data
Ampholytes are compounds which react with both acids and bases. When zinc chloride is reacted with sodium hydroxide the soluble zincate ion is formed. When zinc hydroxide is reacted with hydrochloric acid, the soluble zinc chloride and water are formed.