- Subjects:MS-PS1-2
- Description:Exothermic reactions
- Cat. No.:470338-546
- Includes:Instructional manual and SDS booklet
- No. of uses:5
Study the process of exothermic reactions.
- Understand and demonstrate exothermic reactions
- Aligned to NGSS practices
- Cross cutting concepts: patterns
- Disciplinary core ideas: PS1.A, PS1.B
- Learn engineering practices: analyzing and interpreting data
The reaction is safely carried out in a plastic bag. The students can pass the bag around to feel the result of the reaction.