- Subjects:MS-PS1-2
- Description:Foam city - the catalytic decomposition of hydrogen peroxide
- Cat. No.:470338-542
- Includes:Instructional manual and SDS booklet
- No. of uses:5
A large quantity of foam is produced when detergent and potassium iodide are added to hydrogen peroxide.
- Understand and demonstrate a catalytic decomposition reaction
- Aligned to NGSS practices
- Cross cutting concepts: patterns
- Disciplinary core ideas: PS1.A, PS1.B
- Learn engineering practices: analyzing and interpreting data