Order Entry
United States
Orders LinkContactUsLinkComponent
Formaldehyde 37% (w/w) in aqueous solution stabilised ACS

Formaldehyde 37% (w/w) in aqueous solution stabilised ACS

Plastic bottle, 100 g
Catalog # TS41073-1000
Toxic
Health
Corrosive
Flammable
Supplier #:  41073-1000
Formaldehyde 37% (w/w) in aqueous solution stabilised ACS
Catalog # TS41073-1000
CAS Number:  50-00-0
Formaldehyde 37% (w/w) in aqueous solution stabilised ACS
Formaldehyde 37% (w/w) in aqueous solution stabilised ACS

Plastic bottle, 100 g

CAS Number:  50-00-0
Price
$55.10
Log in for availability
Each (100g)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Synonyms:  Paraformaldehyde aqueous solution, Methanal solution, Methylaldehyde solution, Formol, Formaline solution, Formalin, Methylaldehyde
  • CAS Number:50-00-0
  • Formula:CH₂O
  • MDL Number:MFCD00003274
  • Molecular Weight:30.03 g/mol
  • Melting Pt:–15 °C
  • Flash Pt:50 °C
  • Storage Temperature:Ambient
  • EINECS:200-001-8
  • UN:1198
  • ADR:3,III
  •  :Stabilized with methanol 10-15%
Specifications

Specification Test Results

  • Appearance:
    Clear solution
  • Color scale:
    ≤10 APHA
  • Stabilizer:
    10 to 15 % methanol
  • Residue after ignition:
    ≤0.005 %
  • Titration:
    36.5 to 38.0 %
  • Chloride (Cl):
    ≤5 ppm
  • Sulfate (SO4):
    ≤0.002 %
  • Iron (Fe):
    ≤5 ppm
  • Heavy metals (ICP-OES):
    ≤5 ppm
  • Titrable acid:
    ≤0.006 meq/g

Specifications

  • Pack type:
    Plastic bottle
  • Size:
    100 g

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.