Order Entry
United States
Orders LinkContactUsLinkComponent
 

D-(+)-Mannose 99+%

D-(+)-Mannose 99+%
Certificates
D-(+)-Mannose 99+%

4 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Synonyms:  (2S,3S,4R,5R)-2,3,4,5,6-Pentahydroxyhexanal
  • CAS Number:3458-28-4
  • Formula:C₆H₁₂O₆
  • MDL Number:MFCD00064122
  • Molecular Weight:180.16 g/mol
  • Merck Index:13,05772
  • Melting Pt:133 °C
  • Storage Temperature:Ambient
  • EINECS:222-392-4
Specifications for Product Family

Specification Test Results

  • Appearance:
    White Powder or crystals
  • Infrared spectrum:
    Conforms
  • Loss on drying:
    ≤0.45 %
  • HPLC:
    ≥99.0 %
  • Appearance of solution:
    (10 % in water) Clear
  • Bacterial endotoxins:
    (LAL-test) 0.5 E.U. in 1 ml of a 0.1% soln.
  • Specific optical rotation:
    +13.5° to +14.9° (20°C, 589 nm) (c=10, water) (after 24 hours, on dry substance)
  • Heavy metals (as Pb):
    ≤10 ppm
Product Family Options

Product Information

  • Pack type
    Size
    Availability
    Price
  • Plastic bottle
    25 g
    Log in for availability
    Log in for availability
    Each (25g)
    Each (25g) - $60.40
    $60.40
    Each (25g)
       $60.40

    Supplier Number: 15060-0250
  • Plastic bottle
    1 kg
    Log in for availability
    Log in for availability
    Each (1kg)
    Each (1kg) - $1,090.00
    $1,090.00
    Each (1kg)
       $1,090.00

    Supplier Number: 15060-0010
  • Plastic drum
    2.5 kg
    Log in for availability
    Log in for availability
    Each (2,500g)
    Each (2,500g) - $2,200.00
    $2,200.00
    Each (2,500g)
       $2,200.00

    Supplier Number: 15060-0025
  • Plastic bottle
    100 g
    Log in for availability
    Log in for availability
    Each (100g)
    Each (100g) - $184.00
    $184.00
    Each (100g)
       $184.00

    Supplier Number: 15060-1000

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.