D-(+)-Glucose, anhydrous ACS
Supplier: Thermo Scientific Chemicals
Certificates
About this item
CAS Number:50-99-7
:492-62-6
Formula:C₆H₁₂O₆
MDL Number:MFCD00148912
Merck Index:14,04459
Melting Pt:146 °C
:180.16 g/mol
Storage Temperature:Ambient
EINECS:200-075-1
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specification Test Results
- Appearance:White Crystalline powder
- Infrared spectrum:Conforms
- Loss on drying:≤0.2 % (105°C, 6 h)
- Residue after ignition:≤0.02 %
- Specific optical rotation:+52.5° to +53.0° (25°C, 589 nm) (c=10, H2O, NH4OH)
- Chloride (Cl):≤100 ppm
- SO4 and SO3 (as SO4):≤0.005 %
- Iron (Fe):≤5 ppm
- Heavy metals (as Pb):≤5 ppm
- Titrable acid:≤0.002 meq/g
- Insoluble matter:≤0.005 %
- Starch:Passes test
Specification Test Results
Frequently Bought Together