Order Entry
United States
Orders LinkContactUsLinkComponent
 

D-(-)-Glutamic acid 99+%

D-(-)-Glutamic acid 99+%
Certificates
D-(-)-Glutamic acid 99+%

3 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Synonyms:  (R)-2-Aminopentanedioic acid, (R)-(-)-Glutamic acid, (R)-(-)-2-Aminopentanedioic acid, D(-)-2-Aminopentanedioic acid, D(-)-α-Glutamic acid, D(-)-1-Aminopropane-1,3-dicarboxylic acid, H-D-Glu-OH, (R)-(-)-α-Glutamic acid, (R)-(-)-1-Aminopropane-1,3-dicarboxylic acid
  • CAS Number:6893-26-1
  • Formula:C₅H₉NO₄
  • MDL Number:MFCD00063112
  • Molecular Weight:147.13 g/mol
  • Merck Index:13,04482
  • Boiling Pt:325 °C (1013 hPa)
  • Melting Pt:200…202 °C
  • EINECS:230-000-8
Specifications for Product Family

Specification Test Results

  • Appearance:
    White to off-white Powder
  • Infrared spectrum:
    Conforms
  • Loss on drying:
    ≤0.2 % (105°C, 3 hrs)
  • Sulfated ash:
    ≤0.1 %
  • HPLC:
    ≤1 % L-content
  • Specific optical rotation:
    -30.5° to -32.2° (20°C, 589 nm) (c=10, 2 N HCl)
  • Titration with NaOH:
    99.0 to 101.0 %
  • Other amino acids:
    ≤0.5 % (TLC)
Product Family Options

Product Information

  • Pack type
    Size
    Availability
    Price
  • Glass bottle for solids
    5 g
    Log in for availability
    Log in for availability
    Each (5g)
    Each (5g) - $49.70
    $49.70
    Each (5g)
       $49.70

    Supplier Number: 22789-0050
  • Glass bottle for solids
    25 g
    Log in for availability
    Log in for availability
    Each (25g)
    Each (25g) - $169.00
    $169.00
    Each (25g)
       $169.00

    Supplier Number: 22789-0250
  • Glass bottle for solids
    100 g
    Log in for availability
    Log in for availability
    Each (100g)
    Each (100g) - $477.00
    $477.00
    Each (100g)
       $477.00

    Supplier Number: 22789-1000

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.