

Guanidinium thiocyanate 99+% for molecular biology
Catalog # TS43394-1000
Supplier: Thermo Scientific Chemicals
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Pack type:Glass bottle
- Size:100 g
- Cat. No.:TS43394-1000
Specifications
Specification Test Results
- Appearance:Colorless to white Crystalline powder or crystals
- Infrared spectrum:Conforms
- Melting point:118 to 122 °C
- Titration Argentometric:99.0 to 101.0 %
- Loss on drying:≤0.5 %
- Sulfated ash:≤0.05 %
- Solubility:(3 M in water) Clear colorless
- UV:at 280 nm A: ≤0.4
- pH:5.0 to 7.0 (1 M in water)
- Dnase,Rnase,Protease activity:None detected
- Copper (Cu):≤1 ppm
- Iron (Fe):≤1 ppm
- Lead (Pb):≤1 ppm
- Zinc (Zn):≤2 ppm
Specification Test Results
About this item
:
:118.16 g/mol
Melting Pt:118…122 °C
Storage Temperature:Ambient
UN:3261
ADR:8,III
:GHS07
:GHS05
CAS Number:593-84-0
Formula:CH₅N₃·CHNS
MDL Number:MFCD00013027