Tridecanoic acid 97%
Supplier: Thermo Scientific Chemicals
About this item
Synonyms: n-Tridecanoic acid, Tridecylic acid
- CAS Number:638-53-9
- Formula:C₁₃H₂₆O₂
- MDL Number:MFCD00002741
- Molecular Weight:214.35 g/mol
- Boiling Pt:236 °C (100 mmHg)
- Melting Pt:40…45 °C
- Flash Pt:113 °C
- Storage Temperature:Ambient
- EINECS:211-341-1
3 Options Available Below
Product Details & Documents
Synonyms: n-Tridecanoic acid, Tridecylic acid
- CAS Number:638-53-9
- Formula:C₁₃H₂₆O₂
- MDL Number:MFCD00002741
- Molecular Weight:214.35 g/mol
- Boiling Pt:236 °C (100 mmHg)
- Melting Pt:40…45 °C
- Flash Pt:113 °C
- Storage Temperature:Ambient
- EINECS:211-341-1
Specifications for Product Family
Specification Test Results
- Appearance:White to light yellow Crystalline powder or flakes and/or chunks
- Infrared spectrum:Conforms
- Melting point:40 to 45 °C
- Titration with NaOH:≥96.0 %
Product Family Options
Product Information
- Pack typeSizeAvailabilityPrice
- Supplier Number: 13952-0100
- Supplier Number: 13952-0250
- Supplier Number: 13952-0500
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



