Sulfo-GMBS (N-Maleimidobutyryloxysulfosuccinimide ester), Pierce™
Catalog # PI22324
Supplier: Invitrogen
CAS Number:
Specifications
- Size:50 mg
- Cat. No.:PI22324
- Supplier no.:22324
Specifications
Specification Test Results
- Product Line:Pierce™
- Labeling Method:Chemical Labeling
- Crosslinker Type:Heterobifunctional
- Reactive Moiety:Maleimide, Sulfo-NHS Ester
- Spacer:Short
- Form:Solid
- Quantity:50 mg
- Type:Crosslinker
- Solubility:Water
Specification Test Results
About this item
:
:360.297 g/mol
Storage Temperature:Freezer
CAS Number:158018-86-1
Formula:C₁₂H₁₂N₂O₉S
MDL Number:MFCD00153605