J&W Intuvo Enviromental Specialty Columns, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
Certificates
About this item
Specially designed for analyzing environmental contaminants with the Intuvo GC 9000 system.
- Designed for analysis of environmental semivolatile priority pollutants, pesticides, and volatile organic chemicals
- Tested to ensure consistent analytical results for regulatory methods
- Part of an optimized GC flow path with Agilent Ultra Inert Liners, low-bleed septa, certified vials and gas clean filters
- For use with the Agilent Intuvo 9000 System
Agilent offers various Intuvo GC columns (compatible only with Intuvo 9000 GC System) for the increasingly demanding analysis of environmental contaminants, including pesticides and polycyclic aromatic hydrocarbons (PAH), as well as volatile and semivolatile compounds.
Specifications
- Catalog No:
- AG122-9736-INT
- AGCP7414-INT
- AG123-8336-INT
- AG123-8232-INT
- AG122-1564-INT
- Polarity:
- Mid-polarity stationary phase
- Mid-polarity stationary phase