Benzoylecgonine (CRM), Cayman Chemical
Catalog # 76544-700
Supplier: Cayman Chemical Company
CAS Number:
Specifications
- Concentration:1 mg/ml
- Matrix:Methanol
- Pack type:Glass ampule
- CAS No.:519-09-5
- Description:Benzoylecgonine (CRM)
- Size:1 mg
- Synonym(s):BZE
- Formula:C₁₆H₁₉NO₄
- Storage Temperature:–20 °C
- Description:Benzoylecgonine
- Cat. No.:76544-700
Specifications
About this item
A Certified Reference Material
- This product is qualified as a Certified Reference Material (CRM) that has been manufactured and tested to meet ISO/IEC 17025 and ISO 17034 international standards
- This CRM has been characterized by metrologically valid procedures and may be used as a quantitative analytical reference standard