Δ9-THC (CRM), Cayman Chemical
Supplier: Cayman Chemical Company
Certificates
About this item
A Certified Reference Material
Synonyms:Dronabinol;NSC 134454;Δ9-Tetrahydrocannabinol
Specifications
- CAS No.:1972-08-3
- Formula:C₂₁H₃₀O₂
- Storage Temperature:–20 °C
Specifications
Frequently Bought Together