Δ9-THCB (CRM), Cayman Chemical
Catalog # 76544-586
Supplier: Cayman Chemical Company
CAS Number:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Concentration:1 mg/ml
- Matrix:Acetonitrile
- Pack type:Glass ampule
- CAS No.:60008-00-6
- Description:Δ9-THCB (CRM)
- Size:1 mg
- Synonym(s):∆9-Tetrahydrocannabinol-C4; ∆9-Tetrahydrocannabutol; nor-THC; THCB; ∆9-THC-butyl; ∆9-THC-C4
- Formula:C₂₀H₂₈O₂
- Storage Temperature:–20 °C
- Description:Δ9-THCB
- Cat. No.:76544-586
Specifications
About this item
A Certified Reference Material
- This product is qualified as a Certified Reference Material (CRM) that has been manufactured and tested to meet ISO/IEC 17025 and ISO 17034 international standards
- This CRM has been characterized by metrologically valid procedures and may be used as a quantitative analytical reference standard