Order Entry
United States
ContactUsLinkComponent
9(R)-Δ6a,10a-THC (CRM), Cayman Chemical
9(R)-Δ6a,10a-THC (CRM), Cayman Chemical
Catalog # 76544-576
CAS Number:  
9(R)-Δ6a,10a-THC (CRM), Cayman Chemical
Catalog # 76544-576
Supplier Number:  33013-1
CAS Number:  

Specifications

  • Concentration:
    1 mg/ml
  • Matrix:
    Acetonitrile
  • Pack type:
    Glass ampule
  • CAS No.:
    95720-01-7
  • Description:
    9(R)-Δ6a,10a-THC (CRM)
  • Size:
    1 mg
  • Synonym(s):
    9(R)-Δ6a,10a-Tetrahydrocannabinol; 1(R)-Δ3-THC
  • Formula:
    C₂₁H₃₀O₂
  • Storage Temperature:
    –20 °C
  • Description:
    9(R)-Δ6a,10a-THC
  • Cat. No.:
    76544-576

Specifications

About this item

A Certified Reference Material

  • This product is qualified as a Certified Reference Material (CRM) that has been manufactured and tested to meet ISO/IEC 17025 and ISO 17034 international standards
  • This CRM has been characterized by metrologically valid procedures and may be used as a quantitative analytical reference standard