Order Entry
United States
ContactUsLinkComponent
 
Phytocannabinoid Mixture 11 (CRM) (0.5 ml, 1 mg/ml), Cayman Chemical
  76544-568
 :  
Phytocannabinoid Mixture 11 (CRM) (0.5 ml, 1 mg/ml), Cayman Chemical
  76544-568
 :  32841-1
 :  

 

  • Concentration:
    1 mg/ml of each compound
  • Matrix:
    Acetonitrile
  • Pack type:
    Glass ampule
  • Description:
    Phytocannabinoid Mixture 11 (CRM) (0.5 ml, 1 mg/ml)
  • Size:
    1 mg
  • Storage Temperature:
    –20 °C
  • Cat. No.:
    76544-568

 

 

A mixture of phytocannabinoids in an acetonitrile solution intended for HPLC-UV or LC- or GC-MS

  • This product is qualified as a Certified Reference Material (CRM) that has been manufactured and tested to meet ISO/IEC 17025 and ISO 17034 international standards
  • This CRM has been characterized by metrologically valid procedures and may be used as a quantitative analytical reference standard