Mirtazapine-d3 (CRM), Cayman Chemical
76544-556
:
- Concentration:100 µg/ml
- Matrix:Methanol
- Pack type:Glass ampule
- CAS No.:1216678-68-0
- Description:Mirtazapine-d3 (CRM)
- Size:100 µg
- Synonym(s):6-Azamianserin-d9; ORG 3770-d9
- Formula:C₁₇H₁₆D₃N₃
- Storage Temperature:–20 °C
- Description:Mirtazapine-d3
- Cat. No.:76544-556
A Certified Reference Material
- This product is qualified as a Certified Reference Material (CRM) that has been manufactured and tested to meet ISO/IEC 17025 and ISO 17034 international standards
- This CRM has been characterized by metrologically valid procedures and may be used as a quantitative analytical reference standard