Order Entry
United States
ContactUsLinkComponent
 
MDMB-4en-PINACA (CRM), Cayman Chemical
  76544-532
 :  
MDMB-4en-PINACA (CRM), Cayman Chemical
  76544-532
 :  30761-1
 :  

 

  • Concentration:
    1 mg/ml
  • Matrix:
    Methanol
  • Pack type:
    Glass ampule
  • CAS No.:
    ​2504100-70-1
  • Description:
    MDMB-4en-PINACA (CRM)
  • Size:
    1 mg
  • Synonym(s):
    MDMB-PENINACA; MDMB-PINACA N1-pentyl-4-en isomer
  • Formula:
    C₂₀H₂₇N₃O₃
  • Storage Temperature:
    –20 °C
  • Description:
    MDMB-4en-PINACA
  • Cat. No.:
    76544-532

 

 

A Certified Reference Material

  • This product is qualified as a Certified Reference Material (CRM) that has been manufactured and tested to meet ISO/IEC 17025 and ISO 17034 international standards
  • This CRM has been characterized by metrologically valid procedures and may be used as a quantitative analytical reference standard