Flubromazolam (CRM), Cayman Chemical
76544-422
:
- Concentration:1 mg/ml
- Matrix:Methanol
- Pack type:Glass ampule
- CAS No.:612526-40-6
- Description:Flubromazolam (CRM)
- Size:1 mg
- Formula:C₁₇H₁₂BrFN₄
- Storage Temperature:–20 °C
- Description:Flubromazolam
- Cat. No.:76544-422
A Certified Reference Material
- This product is qualified as a Certified Reference Material (CRM) that has been manufactured and tested to meet ISO/IEC 17025 and ISO 17034 international standards
- This CRM has been characterized by metrologically valid procedures and may be used as a quantitative analytical reference standard