Order Entry
United States
ContactUsLinkComponent
Mephedrone (hydrochloride) (CRM), Cayman Chemical
Mephedrone (hydrochloride) (CRM), Cayman Chemical
Catalog # 76544-404
CAS Number:  
Mephedrone (hydrochloride) (CRM), Cayman Chemical
Catalog # 76544-404
Supplier Number:  26277-1
CAS Number:  

Specifications

  • Concentration:
    1 mg/ml
  • Matrix:
    Methanol
  • Pack type:
    Glass ampule
  • CAS No.:
    1189726-22-4
  • Description:
    Mephedrone (hydrochloride) (CRM)
  • Size:
    1 mg
  • Synonym(s):
    4-MeMC; 4-Methylephedrone; 4-Methylmethcathinone; 4-MMC
  • Formula:
    C₁₁H₁₅NO·HCl
  • Storage Temperature:
    –20 °C
  • Description:
    Mephedrone (hydrochloride)
  • Cat. No.:
    76544-404

Specifications

About this item

A Certified Reference Material

  • This product is qualified as a Certified Reference Material (CRM) that has been manufactured and tested to meet ISO/IEC 17025 and ISO 17034 international standards
  • This CRM has been characterized by metrologically valid procedures and may be used as a quantitative analytical reference standard