Phenyl fentanyl (hydrochloride) (CRM), Cayman Chemical
76544-384
:
- Concentration:100 µg/ml
- Matrix:Methanol
- Pack type:Glass ampule
- Description:Phenyl fentanyl (hydrochloride) (CRM)
- Size:100 µg
- Synonym(s):Benzoyl fentanil; Benzoyl fentanyl; Phenyl fentanil
- Formula:C₂₆H₂₈N₂O·HCl
- Storage Temperature:–20 °C
- Description:Phenyl fentanyl (hydrochloride)
- Cat. No.:76544-384
A Certified Reference Material
- This product is qualified as a Certified Reference Material (CRM) that has been manufactured and tested to meet ISO/IEC 17025 and ISO 17034 international standards
- This CRM has been characterized by metrologically valid procedures and may be used as a quantitative analytical reference standard