Acetaminophen-d4 (CRM), Cayman Chemical
76544-300
:
- Concentration:100 µg/ml
- Matrix:Methanol
- Pack type:Glass ampule
- CAS No.:64315-36-2
- Description:Acetaminophen-d4 (CRM)
- Size:100 µg
- Synonym(s):4-Acetamidophenol-d4; APAP-d4; 4'-Hydroxyacetanilide-d4
- Formula:C₈H₅D₄NO₂
- Storage Temperature:–20 °C
- Description:Acetaminophen-d4
- Cat. No.:76544-300
A Certified Reference Material
- This product is qualified as a Certified Reference Material (CRM) that has been manufactured and tested to meet ISO/IEC 17025 and ISO 17034 international standards
- This CRM has been characterized by metrologically valid procedures and may be used as a quantitative analytical reference standard