Norcarfentanil (hydrochloride) (CRM), Cayman Chemical
Supplier: Cayman Chemical Company
Certificates
About this item
A Certified Reference Material
- This product is qualified as a Certified Reference Material (CRM) that has been manufactured and tested to meet ISO/IEC 17025 and ISO 17034 international standards
- This CRM has been characterized by metrologically valid procedures and may be used as a quantitative analytical reference standard
Specifications
- CAS No.:61085-87-8
- Formula:C₁₆H₂₂N₂O₃·HCl
- Storage Temperature:–20 °C
- Description:Norcarfentanil (hydrochloride)
Specifications
Frequently Bought Together