Order Entry
United States
ContactUsLinkComponent
Midazolam-d4 (maleate) (CRM), Cayman Chemical
Midazolam-d4 (maleate) (CRM), Cayman Chemical
Catalog # 76544-174
CAS Number:  
Midazolam-d4 (maleate) (CRM), Cayman Chemical
Catalog # 76544-174
Supplier Number:  20199-100
CAS Number:  

Specifications

  • Concentration:
    100 µg/ml
  • Matrix:
    Methanol
  • Pack type:
    Glass ampule
  • Description:
    Midazolam-d4 (maleate) (CRM)
  • Size:
    100 µg
  • Formula:
    C₁₈H₉ClD₄FN₃·C₄H₄O₄
  • Storage Temperature:
    –20 °C
  • Description:
    Midazolam-d4 (maleate)
  • Cat. No.:
    76544-174

Specifications

About this item

A Certified Reference Material

  • This product is qualified as a Certified Reference Material (CRM) that has been manufactured and tested to meet ISO/IEC 17025 and ISO 17034 international standards
  • This CRM has been characterized by metrologically valid procedures and may be used as a quantitative analytical reference standard