Ocfentanil-d5 (hydrochloride) (CRM), Cayman Chemical
Catalog # 76544-134
Supplier: Cayman Chemical Company
CAS Number:
Specifications
- Concentration:100 µg/ml
- Matrix:Methanol
- Pack type:Glass ampule
- Description:Ocfentanil-d5 (hydrochloride) (CRM)
- Size:100 µg
- Synonym(s):o-fluoro MAF-d5; o-fluoro Methoxyacetyl fentanyl-d5; ortho-fluoro MAF-d5; ortho-fluoro Methoxyacetyl fentanyl-d5
- Formula:C₂₂H₂₂D₅FN₂O₂·HCl
- Storage Temperature:–20 °C
- Description:Ocfentanil-d5 (hydrochloride)
- Cat. No.:76544-134
Specifications
About this item
A Certified Reference Material
- This product is qualified as a Certified Reference Material (CRM) that has been manufactured and tested to meet ISO/IEC 17025 and ISO 17034 international standards
- This CRM has been characterized by metrologically valid procedures and may be used as a quantitative analytical reference standard