Order Entry
United States
ContactUsLinkComponent
Oxycodone-d6 (CRM), Cayman Chemical
Oxycodone-d6 (CRM), Cayman Chemical
Catalog # 76544-070
CAS Number:  
Oxycodone-d6 (CRM), Cayman Chemical
Catalog # 76544-070
Supplier Number:  19249-1
CAS Number:  

Specifications

  • Concentration:
    1 mg/ml
  • Matrix:
    Methanol
  • Pack type:
    Glass ampule
  • CAS No.:
    152477-91-3
  • Description:
    Oxycodone-d6 (CRM)
  • Size:
    1 mg
  • Synonym(s):
    Dihydro-14-hydroxycodeinone-d6
  • Formula:
    C₁₈H₁₅D₆NO₄
  • Storage Temperature:
    –20 °C
  • Description:
    Oxycodone-d6
  • Cat. No.:
    76544-070

Specifications

About this item

A Certified Reference Material

  • This product is qualified as a Certified Reference Material (CRM) that has been manufactured and tested to meet ISO/IEC 17025 and ISO 17034 international standards
  • This CRM has been characterized by metrologically valid procedures and may be used as a quantitative analytical reference standard